Order Entry
Puerto Rico
ContactUsLinkComponent
 
Louvered bench rack, small, Akro-Mils
  62661-498
 :  Akro-Mils
Louvered bench rack, small, Akro-Mils
  62661-498
 :  Akro-Mils
 :  98600

 

  • Width:
    28"
  • Color:
    Gray
  • Description:
    Freestanding Bench Rack Hanging System
  • Height:
    20"
  • Cat. No.:
    62661-498
  • Supplier no.:
    98-600

 

 

Bench racks keep vital tools and supplies at eye level and within reach.

  • Formed from 16-gauge cold-rolled steel for strength and durability
  • Durable powder-coated finish stands up to heavy-duty service
  • Single-sided
  • Made in the USA

For use with AkroBins or InSight Bins (sold separately).

AkroBins