Order Entry
Puerto Rico
ContactUsLinkComponent
PMSKN Parallel Artificial Membrane Permeability Assay-Skin Kit, BioAssay Systems
PMSKN Parallel Artificial Membrane Permeability Assay-Skin Kit, BioAssay Systems
Catalog # 76545-504
Supplier:  BIOASSAY SYSTEMS
CAS Number:  
undefined
PMSKN Parallel Artificial Membrane Permeability Assay-Skin Kit, BioAssay Systems
Catalog # 76545-504
Supplier:  BIOASSAY SYSTEMS
Supplier Number:  PMSKN-096
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Cat. No.:
    76545-504
  • Details:
    Size: 96 tests

Specifications

About this item

Assess skin membrane permeability of test compounds.

  • Convenient: Includes all necessary equipment to run a PAMPA plate
  • Simple and low-cost: Procedure is easy to follow and more affordable than cell-based permeability assays
  • High-throughput: Can be readily automated as a high-throughput 96-well plate assay for thousands of samples per day
  • Method: PAMPA
  • Samples: Chemical compounds

MEMBRANE PERMEABILITY is an important characteristic to determine for evaluating compounds as potential drug candidates. Drugs often need to cross cell membranes in order to reach their target of action and this makes a compound’s ability to passively cross these membranes an important characteristic to evaluate. The skin, and in particular, the stratum corneum is a complex barrier, which can be mimicked.

Rapid and early screening of compounds for skin penetration is highly desirable for drug discovery. Permeability can be evaluated by cell-based methods; however, these methods are often expensive and time consuming. Parallel Artificial Permeability Assays (PAMPA) offer researchers a quick, inexpensive method of evaluating the permeability of test compounds. Our PMSKN-096 kit is designed to aid in evaluating skin permeability.

BioAssay Systems PMSKN Kit provides all the necessary components to run a Parallel Artificial Permeability Assay for skin permeability studies.