Trypsin Protease, HyClone™
: Cytiva
Dissociation reagents, such as trypsin, are used to detach and disaggregate adherent cells from the surface or substrate to which they are attached, as well as for dissociating tissues for primary cell culture.
- Derived from porcine pancreas
- Gamma irradiated prior to hydration and filling
- Formulated without calcium and magnesium
- Species:Pig
- Storage Conditions:Less than 0 °C
- Enzyme Name:Trypsin
- Formulation:Trypsin, 2.5% 10X Solution, with 25.0 g Porcine Trypsin (1:250/L) in HBSS, without Calcium and Magnesium, without EDTA or Phenol Red