ContactUsLinkComponent
PA53 BIO Trinocular Research Upright 6 MP Digital Microscope Standard Package, Motic
PA53 BIO Trinocular Research Upright 6 MP Digital Microscope Standard Package, Motic
Catalog # 76615-062
Supplier:  Motic
undefined
PA53 BIO Trinocular Research Upright 6 MP Digital Microscope Standard Package, Motic
Catalog # 76615-062
Supplier:  Motic
Supplier Number:  PA53-BIO-1-S6
Specifications
  • Description:
    PA53 BIO upright laboratory microscope
  • Cat. No.:
    76615-062
Specifications
About this item
The PA53 BIO upright laboratory microscopes are designed with the experienced expert in mind. Our PA53 BIO models carry superb optics, ergonomic design and intuitive, automated controls for an efficient workflow, the whole at an unrivalled price/ performance ratio.
  • PA53 BIO trinocular research upright microscope (standard series (w/100 W halogen illumination))
  • 0.5× C-mount camera adpater (PA53 series)
  • Moticam S6
  • Motic analyzes basic 64-bit; USB + dongle (no charge)
The modularity of the PA53 BIO series allows to create tailor-made solutions for your individual needs. Whether it is about color fidelity and extended visual field for pathology, examination of crystals in urine sediments with phase contrast and polarization in urology or advanced FISH/HER2 analysis of chromosomes with fluorescence: The PA53 BIO series covers all of your demands.
Ordering information: Contact your Avantor representative to learn more about this and other Motic solutions. All Motic products come with a one-year electrical and five-year mechanical warranty.