ContactUsLinkComponent
Panthera TEC Polarizing Upright Industrial Microscopes, Motic
Panthera TEC Polarizing Upright Industrial Microscopes, Motic
Catalog # 76557-524
Supplier:  Motic
undefined
Panthera TEC Polarizing Upright Industrial Microscopes, Motic
Catalog # 76557-524
Supplier:  Motic
Supplier Number:  1100104600251
Specifications
  • Description:
    Polarizing binocular
  • Cat. No.:
    76557-524
Specifications
About this item
The panthera TEC POL models present a high flexibility for the inspection of all kind of polarizing materials, for transparent samples as well as for opaque specimen from technical education environments and industrial quality control.
  • UC Plan Achromat objectives (strain-free) for FOV 22 mm
  • Transmitted and transmitted/incident stand options
  • Transmitted light: LED/HAL light source interchangeable
  • Integrated focusable/centerable Bertrand lens
  • Motic lightTracer: Coded nosepiece and digital light intensity knob
The compact Epi illuminator carries a slot for polarizer and analyzer, ready to set up polarization contrast and to reduce internal reflections. An intermediate tube with focusable/centerable Bertrand lens is prepared for a conoscopic analysis of crystals and minerals in transmitted light.
Ordering information: Contact your Avantor representative to learn more about this and other Motic solutions. All Motic products come with a one-year electrical and five-year mechanical warranty.