ContactUsLinkComponent
Corning® BioCoat® Collagen I Cell Culture Inserts, PET Membrane, Corning
  62405-628
 :  Corning
   Low Manufacturing Impact
Corning® BioCoat® Collagen I Cell Culture Inserts, PET Membrane, Corning
  62405-628
 :  Corning
 :  354444
   Low Manufacturing Impact
  • Description:
    24-well
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental Certification:
    ISO 14001 - Bedford (reg. no. 14.4942)
  • Cat. No.:
    62405-628
  • Membrane diameter:
    6.4 mm
  • Membrane material:
    Polyethylene terephthalate (PET)
  • Supplier no.:
    354444
Corning BioCoat cell culture inserts are pre-coated with extracellular matrix proteins for applications requiring a protein-coated cell surface, such as cell adhesion, growth, invasion, migration and/or differentiation.
Coatings include Corning Matrigel® matrix, Fibronectin, Collagen, and Laminin.