InVivoPure™ pH 8.0 Dilution Buffer, BioXCell
About this item
InVivoPure™ dilution buffers are specifically formulated and tested to satisfy the stringent requirements for in vivo applications.
They are extremely low in endotoxin, have been screened for murine pathogens, tested in animal models for toxicity and are formulated with respect to buffer composition and pH to satisfy the requirements of Bio X Cell's antibodies.
This buffer does not contain calcium, magnesium, phenol red, or preservatives such as azide.
Caution: Keep contents sterile. Open only in a biological safety cabinet.
2 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
InVivoPure™ dilution buffers are specifically formulated and tested to satisfy the stringent requirements for in vivo applications.
They are extremely low in endotoxin, have been screened for murine pathogens, tested in animal models for toxicity and are formulated with respect to buffer composition and pH to satisfy the requirements of Bio X Cell's antibodies.
This buffer does not contain calcium, magnesium, phenol red, or preservatives such as azide.
Caution: Keep contents sterile. Open only in a biological safety cabinet.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 76555-584
- 77580-494
- Sterility:
- 0.2 µM filtered
- 0.2 µM filtered
- Shelf Life:
- One year from the date of customer receipt when stored as recommended.
- One year from the date of customer receipt when stored as recommended.
- Storage Temperature:
- 4 °C
- 4 °C
Specifications
Product Family Options
Product Information
- ConcentrationVolumepHAvailabilityPrice
- Supplier Number: IP0080-50ML-Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.76555-584Sterility0.2 µM filteredShelf LifeOne year from the date of customer receipt when stored as recommended.Storage Temperature4 °C - Supplier Number: IP0080-50MLItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.77580-494Sterility0.2 µM filteredShelf LifeOne year from the date of customer receipt when stored as recommended.Storage Temperature4 °C