InVivoPure™ pH 7.0 Dilution Buffer, BioXCell
InVivoPure™ dilution buffers are specifically formulated and tested to satisfy the stringent requirements for in vivo applications.
They are extremely low in endotoxin, have been screened for murine pathogens, tested in animal models for toxicity and are formulated with respect to buffer composition and pH to satisfy the requirements of Bio X Cell's antibodies.
This buffer does not contain calcium, magnesium, phenol red, or preservatives such as azide.
: Keep contents sterile. Open only in a biological safety cabinet.
InVivoPure™ dilution buffers are specifically formulated and tested to satisfy the stringent requirements for in vivo applications.
They are extremely low in endotoxin, have been screened for murine pathogens, tested in animal models for toxicity and are formulated with respect to buffer composition and pH to satisfy the requirements of Bio X Cell's antibodies.
This buffer does not contain calcium, magnesium, phenol red, or preservatives such as azide.
: Keep contents sterile. Open only in a biological safety cabinet.
- Catalog No:
- 76555-692
- 77580-492
- Sterility:
- 0.2 µM filtered
- 0.2 µM filtered
- Shelf Life:
- One year from the date of customer receipt when stored as recommended.
- One year from the date of customer receipt when stored as recommended.
- Storage Temperature:
- 4 °C
- 4 °C
Product Family Options
- ConcentrationVolumepH
- : IP0070-50ML-
Specifications
Cat. No.76555-692Sterility0.2 µM filteredShelf LifeOne year from the date of customer receipt when stored as recommended.Storage Temperature4 °C - : IP0070-50ML
Specifications
Cat. No.77580-492Sterility0.2 µM filteredShelf LifeOne year from the date of customer receipt when stored as recommended.Storage Temperature4 °C