Order Entry
Puerto Rico
ContactUsLinkComponent
Corning® BioCoat® Fibrillar Collagen Cell Culture Inserts, Corning
Corning® BioCoat® Fibrillar Collagen Cell Culture Inserts, Corning
Catalog # 62405-050
Supplier:  Corning
   Low Manufacturing Impact
Corning® BioCoat® Fibrillar Collagen Cell Culture Inserts, Corning
Catalog # 62405-050
Supplier:  Corning
Supplier Number:  354474
   Low Manufacturing Impact

Specifications

  • Description:
    Inserts, 0.1 μm pore size, packed in two 24-well plates
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental Certification:
    ISO 14001 - Bedford (reg. no. 14.4942)
  • Cat. No.:
    62405-050
  • Supplier no.:
    354474

Specifications

About this item

Corning BioCoat cell culture inserts are pre-coated with extracellular matrix proteins for applications requiring a protein-coated cell surface, such as cell adhesion, growth, invasion, migration and/or differentiation.

  • Storage: 2 to 8 °C
  • Shelf life: >= 3 months from date of shipment