Thermo Scientific™ KingFisher Apex Benchtop Sample Prep, Thermo Scientific
Supplier: LIFE TECHNOLOGIES CORP
Certificates
About this item
Benchtop, high-throughput, peak-performance, automated sample extraction and purification system.
- Purify 24 or 96 samples in a single run
- Easy to use interface with touch screen empowering the user to control all details of the application and purification process
- Free BindIx PC software for added convenience
- Extensive selection of validated protocols for different type of applications
- Cloud library for quick protocol retrieval
- Choose from 24, 96 deep-well, and PCR formats, which includes corresponding magnet heads, tip combs, and plates
KingFisher Apex is a versatile and user-friendly instrument which uses magnetic particle processing to automate the purification or isolation of DNA, RNA, proteins, and cells from virtually any source. The Apex moves magnetic beads not liquids through the main steps of sample preparation - bind, wash, elute. This process provides excellent quality, reproducible results, and reduces hands-on time for sample processing.
Specifications
- Application:
- Method / Format:
- Refine target molecule:
- Sample type:
- Target molecule:
- Environmentally Preferable: