ContactUsLinkComponent
Charging stand for three single channel pipettes, Brand
Charging stand for three single channel pipettes, Brand
Catalog # 47751-838
Supplier:  Brandtech
Charging stand for three single channel pipettes, Brand
Catalog # 47751-838
Supplier:  Brandtech
Supplier Number:  705392
Specifications
  • Description:
    Three-instrument charging stand for Transferpette® electronic pipettors, US/Japan plug
  • Cat. No.:
    47751-838
  • Supplier no.:
    705392
Specifications
About this item
Up to 3 single-channel pipettes (up to 1000 µl) can be charged simultaneously in this charging stand.