ContactUsLinkComponent
Eisco Curved Flask Brush
Eisco Curved Flask Brush
  470344-406
 :  Wards
undefined
Eisco Curved Flask Brush
  470344-406
 :  Wards
 :  CH0216B
  • Bristle material:
    Nylon
  • Brush head Ø:
    1.5"
  • Handle material:
    Stainless steel
  • Description:
    Curved flask brush
  • Total length:
    15"
  • Cat. No.:
    470344-406
Designed to scrub the rounded surfaces of flasks, bottles, beakers, cups, jars, dishes, vases and more.
  • Stainless steel, twisted wire handle that is resistant to corrosion
  • Designed for professional laboratories
This semi-stiff, nylon fiber bristle brush is perfect for scrubbing the rounded surfaces of glassware. The stainless steel, twisted handle is semi-flexible, allowing modification to the handle shape as needed, with a looped end for easy hanging. Brush measures 15" in total length. The bristle section measures 6.25" in length and is 1.5" in diameter. Handle measures 8.75" long.