Order Entry
Puerto Rico
ContactUsLinkComponent
1-Pyrimidin-2-ylpiperidin-4-one
Irritant
1-Pyrimidin-2-ylpiperidin-4-one
Catalog # 77405-426
Supplier:  APOLLO SCIENTIFIC
CAS Number:  
Irritant
1-Pyrimidin-2-ylpiperidin-4-one
Catalog # 77405-426
Supplier:  APOLLO SCIENTIFIC
Supplier Number:  OR15509-100MG
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    100 mg
  • Cat. No.:
    77405-426

Specifications

About this item

 :
Melting Pt:73…75 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:116247-92-8
Formula:C₉H₁₁N₃O