Biohazard Waste Bin, Mopec
Product Details & Documents
Mopec’s biohazard waste bin fit the biohazard waste bags perfectly.
- Dimensions: 10×14×19"
- Capacity: 12 to 14 gallons
- Operated by foot pedal for a hands-free lid
- Biohazard waste bags (BG681) fit perfectly inside
- Sold individually
- Does not include bags - BG681 sold separately
Bin features a foot pedal, the touchless lid ensures a quicker, safer way to dispose of hazardous waste.
Specifications
- Description:Biohazard waste bin