Eisco Amber Glass Reagent Bottle with Screw Cap, 100 ml
Catalog # 470344-704
Supplier: Wards
Specifications
- Ø:1.9"
- Capacity:125 ml
- Description:Reagent bottle
- Height:4.6"
- Cat. No.:470344-704
Specifications
About this item
Protects light-sensitive solutions.
- Made of heavy-duty soda glass
- Amber colored glass absorbs UV wavelengths
- Excellent for storing liquids or powders
- Includes white plastic screw cap
Inner diameter of bottle mouth measures 0.7".