Order Entry
Puerto Rico
ContactUsLinkComponent
Eisco Glass Reagent Bottle with Wide Neck and Stopper, 1000 ml
Eisco Glass Reagent Bottle with Wide Neck and Stopper, 1000 ml
  470343-776
 :  Wards
undefined
Eisco Glass Reagent Bottle with Wide Neck and Stopper, 1000 ml
  470343-776
 :  Wards
 :  CH0156FW

 

  • Capacity:
    1000 ml
  • Description:
    Reagent bottle
  • Cat. No.:
    470343-776

 

 

Suitable for mixing and storing samples, liquids, and reagents.

  • Wide neck for easy pouring and decanting
  • Includes a tight fitting glass stopper
  • Made of high quality transparent soda glass - built to last and withstand multiple uses

Cylindrical reagent bottle made of soda glass. Features a wide 5 cm (2") mouth for easy filling and a ground glass stopper seals the bottle to secure contents and help prevent spills and evaporation. This bottle is suitable for use in mixing and storing samples, liquids, and reagents. Size: 8.5"H×4.1"Ø.