Order Entry
Puerto Rico
ContactUsLinkComponent
 
Evergreen Vials, 20 ml, Caplugs Evergreen
  76538-802
 :  Caplugs
undefined
Evergreen Vials, 20 ml, Caplugs Evergreen
  76538-802
 :  Caplugs
 :  240-3519-G80

 

  • Cap material:
    HDPE
  • Capacity:
    20 ml
  • Color:
    Natural
  • Description:
    Vial with writing area
  • Material:
    ok
  • Cat. No.:
    76538-802
  • Cap color:
    Green
  • Bottom style:
    Flat bottom

 

 

Caplugs Evergreen 20 ml Vials are constructed of high-clarity polypropylene copolymer with high density polyethylene screw caps.

  • Externally molded-in graduations in 3 ml intervals
  • Molded-in frosted writing area for easy labeling
  • Specially designed tray helps eliminate accidental knockovers and expedites processing
  • Vial has an octagonal-shaped base which locks into the alpha-numerically labeled tray to allow for easy one-handed capping and uncapping

These vials have a wide mouth opening for ease of use and the clarity of polypropylene material provides for clear visibility of the contents.