ContactUsLinkComponent
Klercide™ Neutral Detergent Unit Dose Concentrate
Klercide™ Neutral Detergent Unit Dose Concentrate
Catalog # 76554-262
undefined
Klercide™ Neutral Detergent Unit Dose Concentrate
Catalog # 76554-262
Supplier Number:  6000186
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Size:
    50x100 mL
  • Cat. No.:
    76554-262
  • Packaging:
    Tub, double bagged
  • Sterility:
    Sterile
  • Form:
    Ready-to-use
Specifications
About this item
Klercide™ Neutral Detergent is a low foaming non-ionic detergent solution that is a reliable residue cleaner formulated for cleanrooms.
  • Sterile Neutral Formulation - pH-neutral, non-ionic, non-corrosive, low-foaming detergent — with no extraneous chemicals, dyes or perfumes — is designed to be used directly on hard, nonporous surfaces.
  • Powerful Cleaning Performance - Designed specifically for use in cleanrooms and other controlled areas, Klercide™ Neutral Detergent is highly effective against oils, sticky residues and spillages, and ideal for use in general cleaning protocols.
  • Simple, Consistent Unit Dose Format - Pre-measured unit-dose concentrate containers streamline dilution, ensuring consistency and efficiency while minimizing operator exposure risks. Containers are double-bagged to facilitate movement into high-grade areas.
  • Cleanroom-Grade Sterile Production - Unit dose concentrate is 0.2 micron-filtered and filled under unidirectional Grade A / ISO Class 7 airflow in a Grade B / ISO Class 5 cleanroom — and double-bagged to simplify movement into high-grade areas. Supported with Certificates of Analysis and Sterility with every batch.
Ecolab Klercide™ Neutral Detergent is 0.2 micron filtered then filled and bagged in a Grade C / ISO Class 5 cleanroom before being terminally sterilized by gamma irradiation. The product is a sterile detergent concentrate, to be diluted at a 1:49 ratio, and ideal for large surfaces.