ContactUsLinkComponent
Klercide™ Low Residue Peroxide
Klercide™ Low Residue Peroxide
Catalog # 76554-252
undefined
Klercide™ Low Residue Peroxide
Catalog # 76554-252
Supplier Number:  6000187
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Size:
    6 x 32oz.
  • Cat. No.:
    76554-252
  • Packaging:
    Trigger spray
  • Sterility:
    Sterile
  • Form:
    Ready-to-use
Specifications
About this item
Klercide™ low residue peroxide is a sterile hydrogen peroxide solution with low residues designed for pharmaceutical cleanrooms.
  • Powerful cleaning and residue removal / designed specifically for use in cleanrooms and other controlled areas where a low-residue product is required, providing an effective alternative to IPA in general cleaning protocols.
  • Low residue, no-rinse use - low-residue formulation enables highly effective cleaning and reside removal / without the need to wipe down with alcohol or water afterwards / streamlining cleaning protocols.
  • Ready-to-use formats - available in trigger spray and capped bottle formats / all designed to reduce handling and need for dilution, to streamline cleaning protocols and reduce cost-to-clean.
  • Sterile delivery system - trigger-spray format features patented SteriShield™ delivery system (SDS), providing sterility assurance of contents in bottle by forming a complete seal that prevents any air from (microbial or chemical contaminants) being drawn back into the bottle.
  • Cleanroom-grade sterile production - solution is 0.2 micron-filtered and filled under unidirectional Grade A/ISO Class 7 airflow in a Grade B/ISO Class 5 cleanroom / and double-bagged to simplify movement into high-grade areas. Supported with Certificates of analysis and sterility with every batch.
For use in cleaning and removing residues, Klercide™ Low Residue Peroxide is suitable for use on all types of hard surfaces in cleanrooms, controlled environments, and isolators. The product is a blend of high grade 6% hydrogen peroxide and Water For Injection (WFI).
Klercide low residue peroxide is a 0.2 micron filtered, bagged and terminally sterilized by gamma irradiation. Endotoxin tested to an approved pharmacopeia method.