Klercide™ Cleanroom Quat Metered Dose Concentrate Sterile
76554-250
: Avantor
- Size:4x1 gal
- Cat. No.:76554-250
- Packaging:Bottle, double bagged, capped
- Sterility:Sterile
- Form:Metered dose concentrate
Klercide™ Cleanroom Quat Metered Dose Concentrate Sterile is a blend of quaternary ammonium compound blended with Water for Injection. Concentrated non-food contact sanitizer, disinfectant, virucide, and fungicide for hard, non-porous surfaces for use in cleanrooms and controlled areas.
- Formulated to leave minimal visible residues when used as a non-food contact sanitizer on hard, non-porous surfaces
- Is easy and economical to use
- Is 0.2 micron filtered, filled, and double bagged in a cleanroom environment
- Compatible with most surfaces found in a cleanroom
- Compatible with other Ecolab biocides for rotational use
Each 1-gallon container makes 50 gallons of diluted sanitizer (76 ml of concentrate makes 1 gallon of ready-to-use sanitizer). Flexible use instructions allow for product use as a sanitizer, disinfectant, or fungicidal application.
EPA Registered Sanitizer, Disinfectant, Virucide and Fungicide.
EPA Reg. # 1677-264