ContactUsLinkComponent
Klercideā„¢ 70/30 IPA - Canada
Klercideā„¢ 70/30 IPA - Canada
Catalog # 76554-228
undefined
Klercideā„¢ 70/30 IPA - Canada
Catalog # 76554-228
Supplier Number:  6600140
Specifications
  • Size:
    6x32 oz
  • Cat. No.:
    76554-228
  • Packaging:
    Trigger spray bottle, capped
  • Sterility:
    Sterile
  • Form:
    Ready-to-use
Specifications
About this item
Klercideā„¢ 70/30 IPA is sterile 70% v/v Isopropyl alcohol blended with Water for Injection for daily disinfection of critical areas. Klercideā„¢ 70/30 IPA is a one-step disinfectant IPA spray, designed for disinfection of cleanroom environments, with a trigger-spray format featuring the patented ā€œbag-in-a-bottleā€ SteriShieldā„¢ Delivery System.
  • Efficacy - 0.2 micron-filtered 70% isopropyl alcohol solution blended with WFI (endotoxin level guaranteed to be <0.25E.U./mL>) to deliver Bactericidal, Fungicidal and Virucidal efficacy.
  • Ideal for Sanitizing - Our 70/30 IPA is ideal for sanitizing hard, non-porous, food contact and non-food contact surfaces.
  • Daily Disinfection - 70/30 IPA in a user-friendly trigger-spray format, designed to streamline daily disinfection in critical processing applications including aseptic processing, RABs, product contact surfaces and other sterile areas
  • Sterile Delivery System - Trigger-spray format features patented SteriShieldā„¢ Delivery System (SDS), providing sterility assurance of contents in bottle by forming a complete seal that prevents any air from (and microbial or chemical contaminants) being drawn back into the bottle.
  • Sterile Production - Ultrasonically sealed and gamma irradiated in a Grade C/ISO Class 5 cleanroom; supported with certificates of analysis and Certificates of Sterility with every batch.
  • Convenient Packaging - Available as either double or triple-bagged to streamline movement into high-grade areas, with packaging designed for ease-of-use with gloved hands.
For use in pharmaceutical cleanrooms and associated areas, Klercideā„¢ 70/30 IPA can be used for disinfection purposes on cleanroom surfaces and equipment. Available in a ready-to-use format, Klercide 70/30 IPA contains 70% v/v Isopropyl Alcohol blended with Water for Injection.
The product is 0.2 micron filtered and bagged in an ISO 5 cleanroom before being terminally sterilized by gamma irradiation. Endotoxin level is guaranteed to be less than 0.25EU/ml.
Health Canada DIN registered Disinfectant, Sanitizer, Fungicide and Virucide.
DIN 02447789