COSA™ PUR 85 oxidizing additive
Supplier: ECOLAB LIFE SCIENCES
Certificates
About this item
COSA™ PUR 85 is a liquid oxidizing additive.
- Helps in adhering to wastewater permissible limits.
- Promotes Quality Assurance
- In combination with alkaline cleaners, COSA PUR 85 enhances the removal of stubborn soiling
- Excellent foam suppressing properties
- Excellent soil loading properties
- COSA PUR 85 is supported by a complete validation package including techniques for determining acceptable residual levels and analytical methods for residue quantitation
COSA™ PUR 85 is a liquid oxidizing additive for alkaline cleaners to enhance cleaning in the pharmaceutical and personal care industries.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.