ContactUsLinkComponent
Haug Anti-Static Brush, Mettler-Toledo
Haug Anti-Static Brush, Mettler-Toledo
Catalog # 76580-228
Supplier:  Mettler Toledo
Haug Anti-Static Brush, Mettler-Toledo
Catalog # 76580-228
Supplier:  Mettler Toledo
Supplier Number:  63052301
Specifications
  • Description:
    Brush, Haug anti-static
  • Cat. No.:
    76580-228
Specifications
About this item
Anti-static brush to remove electrostatic charges, prevent cross contanimation, and enable accurate weighing results.