RheoPacs, Port Tops, Rheo Flexibles (Division of Rheo Engineering)
Supplier: RHEO ENGINEERING LLC
Certificates
About this item
Rheo Flexibles provides a wide range of single-use transfer containers. The RheoPac family of products offers an excellent alternative to stainless intermediate transfer containers, and can be configured with many volumes and port options.
- Constructed from RheoFlex 302 polyethylene material
- FDA approved for packaging
- USP and EP compliant
- 21 CFR 177.1520 compliant
- Matte finish
- Anti-static film
Designed with integrated HDPE ports and handling loops to allow easy filling, transporting, and unloading.
Certifications: USP <661.1>, USP <661.2>, USP <88>, USP <87>, USP Class VI, EP 3.1.3, EU 10/2011, 94/62/EC, CONEG.
Caution: Store in box, out of UV light, in a cool, dry location.