Order Entry
Puerto Rico
ContactUsLinkComponent
VWR® Standard Hazardous Location (Explosion Proof) Laboratory Refrigerator with Manual Defrost (30 cu. ft.)
VWR® Standard Hazardous Location (Explosion Proof) Laboratory Refrigerator with Manual Defrost (30 cu. ft.)
Catalog # 76580-132
Supplier:  VWR International
undefined
VWR® Standard Hazardous Location (Explosion Proof) Laboratory Refrigerator with Manual Defrost (30 cu. ft.)
Catalog # 76580-132
Supplier:  VWR International

Specifications

  • Temperature range:
    1...10 °C
  • Capacity:
    850 L (30 cu.ft.)
  • Description:
    Standard series laboratory refrigerator upright hazardous location (explosion proof), 30 CF
  • Cat. No.:
    76580-132
  • Electrical:
    115 V, 60 Hz, 6 A, ⅓ HP
  • Defrost:
    Cycle
  • Door Type:
    1 solid door, right hinged
  • Shelves/drawers:
    4 fixed shelves
  • Weight:
    178 kg (393 lbs.)
  • Exterior W×D×H:
    908×889×1969 mm (35¾×35×77½")
  • Interior W×D×H:
    813×572×1651 mm (32×22½×65")

Specifications

About this item

They utilize non-sparking and/or ATEX rated components and are engineered for use in environments where volatile/explosive conditions could potentially exist. For storage of flammable or volatile materials within typical laboratory or non-volatile environments, see flammable material storage units.

  • Mechanical thermostat sealed in explosion proof box
  • Keyed door lock
  • High density foam insulation
  • Must be hardwired for safety

These units are equipped with interior shelving, and microprocessor temperature controllers. The standard line provides multi-functional features in a cost-effective design.

Material: White powder coat

Certifications: These units meet NFPA and OSHA guidelines for hazardous location, Class 1, Division II, Groups C and D guidelines.
Ordering information: Units come with a two year parts and labor warranty.
Caution: Units are not equipped with a line cord and plug. They must be hardwired in conduit to 120 VAC power source for safety.