VWR® Standard Hazardous Location (Explosion Proof) Laboratory Freezer with Manual Defrost (30 cu. ft.)
Catalog # 76580-128
Supplier: VWR International
Specifications
- Temperature range:‒15...‒25 °C
- Capacity:850 L (30 cu.ft.)
- Description:Standard series laboratory freezer upright hazardous location (explosion proof), 30 CF
- Cat. No.:76580-128
- Electrical:115 V, 60 Hz, 6 A, ⅓ HP
- Defrost:Manual
- Door Type:1 solid door, right hinged
- Shelves/drawers:4 fixed shelves
- Weight:177 kg (390 lbs.)
- Exterior W×D×H:908×889×1969 mm (35¾×35×77½")
- Interior W×D×H:813×572×1651 mm (32×22½×65")
Specifications
About this item
They utilize non-sparking and/or ATEX rated components and are engineered for use in environments where volatile/explosive conditions could potentially exist. For storage of flammable or volatile materials within typical laboratory or non-volatile environments, see flammable material storage units.
- Mechanical thermostat sealed in explosion proof box
- Keyed door lock
- High density foam insulation
- Must be hardwired for safety
These units are equipped with interior shelving, and microprocessor temperature controllers. The standard line provides multi-functional features in a cost-effective design.
Material: White powder coat
Certifications: These units meet NFPA and OSHA guidelines for hazardous location, Class 1, Division II, Groups C and D guidelines.
Ordering information: Units come with a two year parts and labor warranty.
Caution: Units are not equipped with a line cord and plug. They must be hardwired in conduit to 120 VAC power source for safety.