Order Entry
Puerto Rico
ContactUsLinkComponent
 
SARS-CoV-2 (2019-nCoV) Recombinant Spike RBD(A372S)-His Protein (from HEK293 Cells)
  103886-074
 :  
SARS-CoV-2 (2019-nCoV) Recombinant Spike RBD(A372S)-His Protein (from HEK293 Cells)
  103886-074
 :  40592-V08H19
 :  

 

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    SARS-CoV-2
  • Size:
    100 µg
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    <1.0 EU per μg protein as determined by the LAL method.
  • Gene ID:
    YP_009724390.1
  • Reconstitution Instructions:
    A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    ncov RBD|ncov s2|ncov spike|coronavirus s2|S|s1|Spike RBD|ncov s1|coronavirus spike|coronavirus s1|cov spike|RBD
  • Protein/Peptide Name:
    Spike
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    The recombinant SARS-CoV-2 (2019-nCoV) Spike RBD(A372S)-His Recombinant Protein consists of 234 amino acids and predicts a molecular mass of 26.6 kDa.
  • Sequence:
    Arg319-Phe541(A372S)
  • Shipping Temperature:
    Ambient
  • Cat. No.:
    103886-074

 

 

A DNA sequence encoding the SARS-CoV-2 (2019-nCoV) Spike RBD(A372S)-His Recombinant Protein (YP_009724390.1) (Arg319-Phe541(A372S)) was expressed with a polyhistidine tag at the C-terminus.