SARS-CoV-2 (B.1.621) Recombinant S1+S2 trimer Protein (ECD, His Tag) (HPLC-verified) (from HEK293 Cells)
Catalog # 76535-094
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:SARS-CoV-2
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg protein as determined by the LAL method.
- Biological Activity:Immobilized human ACE2 protein (mFc tag)(Cat:10108-H05H) at 2 μg/ml (100 μl/well) can bind SARS-CoV-2 (B.1.621) S1+S2 trimer Protein ( ECD, His Tag)(Cat:40589-V08H20), the EC50 is 2-20 ng/ml.
- Gene ID:YP_009724390.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:ncov RBD|ncov s2|ncov spike|coronavirus s2|S|s1|Spike RBD|ncov s1|coronavirus spike|coronavirus s1|cov spike|RBD
- Protein/Peptide Name:Spike
- Purity:>95% as determined by SDS-PAGE. >90% as determined by SEC-HPLC.
- Molecular Weight:The recombinant SARS-CoV-2 Spike S1+S2 consists of 1231 amino acids and predicts a molecular mass of 136.41 kDa.
- Sequence:Met1-Gln1208
- Shipping Temperature:Ambient
- Cat. No.:76535-094
Specifications
About this item
A DNA sequence encoding the SARS-CoV-2 Spike S1+S2 (YP_009724390.1, with mutations T95I, Y144S, Y145N, R346K, E484K, N501Y, D614G, P681H, F817P, A892P, A899P, A942P, D950N, K986P, V987P and furin cleavage site mutants) expressed with the bacteriophage T4 fibritin and a polyhistidine tag at the C-terminus. The mutations were identified in the SARS-CoV-2 variant (known as variant B.1.621) which emerged in the South America.