SARS-CoV-2 (AY.4.2) Recombinant Spike S1+S2 Protein (ECD, His Tag) (from Baculovirus Insect Cells)
Catalog # 76535-088
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:SARS-CoV-2
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg protein as determined by the LAL method
- Biological Activity:Immobilized human ACE2 protein (mFc tag)(Cat:10108-H05H) at 2 μg/ml (100 μl/well) can bind Recombinant SARS-CoV-2 (BA.4/BA.5/BA.5.2) Spike S1+S2 trimer Protein (ECD, His Tag)(Cat:40589-V08H32), the EC50 is 8-32 ng/ml.
- Gene ID:YP_009724390.1
- Reconstitution Instructions:A hardcopy of COA with reconstitution instruction is sent along with the products, please refer to it for detailed information
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:ncov RBD|ncov s2|ncov spike|coronavirus s2|S|s1|Spike RBD|ncov s1|coronavirus spike|coronavirus s1|cov spike|RBD
- Protein/Peptide Name:Spike
- Purity:>90% as determined by SDS-PAGE, >90% as determined by SEC-HPLC
- Molecular Weight:The recombinant SARS-CoV-2 (BA.4/BA.5/BA.5.2) Spike S1+S2 consists of 1226 amino acids and predicts a molecular mass of 136.26 kDa.
- Sequence:Met1-Gln1208
- Formulation:Lyophilized from sterile 25 mM sodium citrate, 200 mM NaCl, 0.02% tween80, 10% Trehalose, pH 6.0
- Shipping Temperature:Ambient
- Cat. No.:76535-088
Specifications
About this item
A DNA sequence encoding the SARS-CoV-2 Spike S1+S2 (YP_009724390.1, with mutations T19R, E156G, 157-158 deletion, A222V, L452R, T478K, D614G, P681R, D950N) (Met1-Pro1213) was expressed with a polyhistidine tag at the C-terminus.