SARS-CoV-2 Recombinant Spike S1+S2 (T19R, E156G, 157-158 deletion, L452R, T478K, D614G, P681R, D950N) Protein (ECD, His Tag) (from Baculovirus Insect Cells)
76535-086
:
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Baculovirus (Insect cells)
- Species:SARS-CoV-2
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg protein as determined by the LAL method.
- Biological Activity:Immobilized ACE2 Protein, Human, Recombinant (mFc Tag)(Cat:10108-H05H) at 2 μg/ml (100 μl/well) can bind SARS-CoV-2 Spike S1+S2 (T19R, E156G, 157-158 deletion, L452R, T478K, D614G, P681R, D950N) Protein (ECD, His Tag)(Cat:40589-V08B27), the EC50 is 200-1200 ng/ml.
- Gene ID:YP_009724390.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:ncov RBD|ncov s2|ncov spike|coronavirus s2|S|s1|Spike RBD|ncov s1|coronavirus spike|coronavirus s1|cov spike|RBD
- Protein/Peptide Name:Spike
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:The recombinant SARS-CoV-2 Spike S1+S2 consists of 1207 amino acids and predicts a molecular mass of 134.14 kDa.
- Sequence:Met1-Pro1213
- Shipping Temperature:Ambient
- Cat. No.:76535-086
A DNA sequence encoding the SARS-CoV-2 Spike S1+S2 (YP_009724390.1, with mutations T19R, E156G, 157-158 deletion, L452R, T478K, D614G, P681R, D950N) (Met1-Pro1213) was expressed with a polyhistidine tag at the C-terminus. The mutations were identified in the SARS-CoV-2 variant (known as variant AY.3, AY.4) which emerged in the India.