Human Recombinant LILRB5/CD85c/LIR-8 Protein (ECD, Fc Tag) (from HEK293 Cells)
76537-318
:
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg protein as determined by the LAL method.
- Gene ID:NP_006831.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:LIR-8|LIR8|CD85C
- Protein/Peptide Name:LILRB5
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:The recombinant human LILRB5 consists of 673 amino acids and predicts a molecular mass of 74.06 kDa. It migrates as an approximately 91.35 kDa band in SDS-PAGE under reducing conditions.
- Sequence:Met1-Gly458
- Shipping Temperature:Ambient
- Cat. No.:76537-318
A DNA sequence encoding the human LILRB5 (NP_006831.1) (Met1-Gly458) was expressed with the Fc region of human IgG1 at the C-terminus.