Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant LILRB4/CD85k/ILT3 Protein (ECD,His Tag) (from HEK293 Cells)
Human Recombinant LILRB4/CD85k/ILT3 Protein (ECD,His Tag) (from HEK293 Cells)
Supplier:  Sino Biological
Certificates
Human Recombinant LILRB4/CD85k/ILT3 Protein (ECD,His Tag) (from HEK293 Cells)
Supplier:  Sino Biological

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    <1.0 EU per μg protein as determined by the LAL method.
  • Gene ID:
    NP_001265357.2
  • Reconstitution Instructions:
    A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    CD85K|ILT-3|ILT3|LIR5|LIR-5
  • Protein/Peptide Name:
    LILRB4
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    The recombinant human LILRB4 consists of 249 amino acids and predicts a molecular mass of 27.7 kDa. As a result of glycosylation, it migrates as an approximately 35 - 42 KDa band in SDS-PAGE under reducing conditions.
  • Sequence:
    Met1-Glu259
  • Shipping Temperature:
    Ambient

Specifications