Order Entry
Puerto Rico
ContactUsLinkComponent
 
Human Recombinant CD30/TNFRSF8 Protein (His & AVI Tag), Biotinylated (from HEK293 Cells)
 
undefined
Human Recombinant CD30/TNFRSF8 Protein (His & AVI Tag), Biotinylated (from HEK293 Cells)

 

  • Conjugation:
    Biotin
  • Protein Function:
    Immunotherapy
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage Conditions:
    −20…−80 °C
  • Endotoxin Content:
    <1.0 EU per μg protein as determined by the LAL method.
  • Gene ID:
    NP_001234.2
  • Reconstitution Instructions:
    A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    Ki-1|D1S166E|Cd30|Ki
  • Protein/Peptide Name:
    TNFRSF8
  • Purity:
    >95% as determined by SDS-PAGE
  • Molecular Weight:
    The recombinant human TNFRSF8 consists of 387 amino acids and predicts a molecular mass of 41.74 kDa. It migrates as an approximately 78.49 kDa band in SDS-PAGE under reducing conditions.
  • Sequence:
    Met1-Lys379
  • Shipping Temperature:
    Ambient