VWR® Autoclavable Cleanroom Goggles
Catalog # 76579-452
Supplier: VWR International
Specifications
- Frame color:White
- Description:Cleanroom goggles
- Environmentally Preferable:
- Cat. No.:76579-452
- Lens coating:Anti-fog, anti-scratch
- Lens tint:Clear
Specifications
About this item
Cleanroom goggles feature a white frame with indirect ventilation.
- Anti-fog and anti-scratch protection
- Clear lens
- Indirect ventilation
- Silicone strap
- White frame
- Autoclavable
- Tested to withstand >40 autoclave cycles (30 minutes duration at 121/250 °C)
Clear replacement lenses also feature anti-fog and anti-scratch protection.
Ordering information: Replacement lenses are sold separately.