Order Entry
Puerto Rico
ContactUsLinkComponent
Blue Sepharose™ High Performance Affinity Chromatography Media, Cytiva
Blue Sepharose™ High Performance Affinity Chromatography Media, Cytiva
Catalog # 104013-192
Supplier:  Cytiva
CAS Number:  
Blue Sepharose™ High Performance Affinity Chromatography Media, Cytiva
Catalog # 104013-192
Supplier:  Cytiva
Supplier Number:  90100123
CAS Number:  

Specifications

  • Particle Size:
    ~34 µm
  • Description:
    Blue Sepharose™ High Performance
  • pH Range:
    4 - 12
  • Matrix:
    Highly cross-linked agarose, 6%
  • Ligand:
    Cibacron Blue F3G-A
  • Storage Temperature:
    4 - 30 °C
  • Ligand Density:
    4 mg Cibacron Blue F3G-A/ml medium
  • Ligand Coupling Method:
    Triazine
  • Cat. No.:
    104013-192
  • Type:
    Liquid

Specifications

About this item

This media is suitable for affinity chromatography.

Chemical stability: Commonly used buffers, 70% ethanol, 8 M urea and 6 M guanidine hydrochloride.

Storage solution: 20% ethanol