Cynomolgus Recombinant IL12RB2 Protein (mFc Tag) (from HEK293 Cells)
A DNA sequence encoding the cynomolgus IL12RB2 (XP_005543136.1) (Lys24-Asn623) was expressed with the Fc region of mouse IgG2a at the C-terminus.
- Conjugation:Unconjugated
- Protein Function:Immunotherapy
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Cynomolgus monkey
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg protein as determined by the LAL method.
- Gene ID:XP_005543136.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:IL-12RB2|A930027I18Rik|Ifnm
- Protein/Peptide Name:IL12RB2
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:The recombinant Cynomolgus IL12RB2 consists of 839 amino acids and predicts a molecular mass of 94.93 kDa. It migrates as an approximately 111.51 kDa band in SDS-PAGE under reducing conditions.
- Sequence:Lys24-Asn623
- Shipping Temperature:Ambient