Mouse Recombinant FGL1 Protein (His Tag) (from Baculovirus Insect Cells)
Catalog # 76537-394
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Baculovirus (Insect cells)
- Species:Mouse
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg protein as determined by the LAL method.
- Gene ID:NP_663569.2
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:HP-041|HFREP1|LFIRE1|LFIRE-1
- Protein/Peptide Name:FGL1
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:The recombinant mouse FGL1 consists of 303 amino acids and predicts a molecular mass of 35.5 kDa.
- Sequence:Met1-Ile314
- Shipping Temperature:Ambient
- Cat. No.:76537-394
Specifications
About this item
A DNA sequence encoding the mouse FGL1 (NP_663569.2) (Met1-Ile314) was expressed with a polyhistidine tag at the C-terminus.
- Shipping temperature: Dry ice