Human Recombinant respiratory syncytial virus (RSV) Nucleoprotein Protein (His Tag) (from E. coli)
76535-114
:
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:RSV
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:Please contact us for more information.
- Gene ID:Q6V2F2
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:RSV Nucleoprotein
- Protein/Peptide Name:RSV-NP
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:The recombinant Human respiratory syncytial virus (RSV) Nucleoprotein consists of 398 amino acids and predicts a molecular mass of 44.39 kDa. It migrates as an approximately 39.79 kDa band in SDS-PAGE under reducing conditions.
- Sequence:Met1-Leu391
- Shipping Temperature:Ambient
- Cat. No.:76535-114
A DNA sequence encoding the human respiratory syncytial virus (RSV) nucleoprotein (Q6V2F2) (Met1-Leu391) was expressed with a polyhistidine tag at the N-terminus.