Bovine High Density LipoProtein (HDL) ELISA Kit, AFG Bioscience
Catalog # 76578-336
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Host:Bovine
- Target protein:High density lipoprotein
- Description:Bovine High density lipoprotein(HDL) ELISA kit
- Size:96 Wells
- Sample Type:Serum, plasma, urine, tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:—
- Storage Temperature:2…8 °C
- Sample Volume:50 - 100 µl
- Sensitivity:—
- Regulatory Status:RUO
- Cat. No.:76578-336
Specifications
About this item
- Research Topic: Cardiovascular
- Abbreviation: HDL
- Detection Wavelength: 450 nm