Bovine Ectonucleoside Triphosphate Diphosphohydrolase 1 (ENTPD1) ELISA Kit, AFG Bioscience
Catalog # 76578-564
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Host:Bovine
- Target protein:Ectonucleoside triphosphate diphosphohydrolase 1
- Description:Bovine Ectonucleoside triphosphate diphosphohydrolase 1(ENTPD1) ELISA kit
- Size:96 Wells
- Sample Type:Serum, plasma, urine, tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:—
- Storage Temperature:2…8 °C
- Sample Volume:50 - 100 µl
- Sensitivity:—
- Regulatory Status:RUO
- Cat. No.:76578-564
Specifications
About this item
- Research Topic: Metabolism
- Abbreviation: ENTPD1
- Detection Wavelength: 450 nm