Mouse cross linked N-TeloPeptide of type Ⅰ collagen (NTX) ELISA Kit, AFG Bioscience
Catalog # 76578-690
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Host:Mouse
- Target protein:Cross linked N-telopeptide of type Ⅰ collagen,NTX
- Description:Mouse cross linked N-telopeptide of type Ⅰ collagen,NTX ELISA kit
- Size:96 Wells
- Sample Type:Serum, plasma, urine, tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:0.6 - 40 ng/ml
- Storage Temperature:2…8 °C
- Sample Volume:50 - 100 µl
- Sensitivity:0.1 ng/ml
- Regulatory Status:RUO
- Cat. No.:76578-690
Specifications
About this item
- Assay Principle: Quantitative
- Research Topic: Cancer ,Cell Biology
- Detection Wavelength: 450 nm