Fixative Dissect Aid™, Statlab
Supplier: StatLab
Jump to:
About this item
Dissect-aid is a fixative which helps make lymph nodes more identifiable during pathology examination.
- The product turns lymph nodes pale/white so they stand out from other tissue
- Solution firms the nodes for touch identification when performing gross examination
- Dramatically improves lymph node recovery/harvesting, and also an excellent fixative for fatty tissues
- This formulation has helped in diagnosis for the past 30 years
Part of the StatLab line of prefilled bulk containers.
Manufactured in the USA
2 Options Available Below
Product Details & Documents
Dissect-aid is a fixative which helps make lymph nodes more identifiable during pathology examination.
- The product turns lymph nodes pale/white so they stand out from other tissue
- Solution firms the nodes for touch identification when performing gross examination
- Dramatically improves lymph node recovery/harvesting, and also an excellent fixative for fatty tissues
- This formulation has helped in diagnosis for the past 30 years
Part of the StatLab line of prefilled bulk containers.
Manufactured in the USA