Specifications
- Description:Fixative StatFix™
- Size:1 gallon
- Cat. No.:76548-204
Specifications
About this item
Buffered alcoholic formalin fixative.
- Particularly well suited for fatty tissues, rapid diagnostic applications and routine frozen sections
Part of the StatLab line of prefilled bulk containers.
Manufactured in the USA