ContactUsLinkComponent
Formalin Substitute GTF™, Statlab
  76548-114
 :  StatLab
 :  
undefined
Formalin Substitute GTF™, Statlab
  76548-114
 :  StatLab
 :  FS-1
 :  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    Formalin Substitute GTF™
  • Size:
    1 gallon
  • Cat. No.:
    76548-114
A glyoxyl based, formalin-free fixative.
  • Excellent for conventional histology, special stains and immunohistochemistry
  • The formaldehyde-free solution provides the morphological detail of a standard aldehyde fixative while preserving high levels of immunoreactivity
Part of the StatLab line of formalin alternatives.
Manufactured in the USA