Zinc Formalin Buffered , Statlab
Supplier: StatLab
Certificates
About this item
Ready-to-use Zinc Formalin solution.
- Especially recommended for use in immunohistochemistry
- Zinc formalin preserves more immunoreactivity than standard NBF
Part of the StatLab line of StatClick 10% NBF Prefills.
Manufactured in the USA
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.