Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Xylene Substitute XS-3™, Statlab

Xylene Substitute XS-3™, Statlab

Xylene Substitute XS-3™, 1 gallon
Catalog # 76548-058
Supplier #:  7400-1
Xylene Substitute XS-3™, Statlab
Catalog # 76548-058
Supplier:  
CAS Number:  
Xylene Substitute XS-3™, Statlab
Xylene Substitute XS-3™, Statlab

Xylene Substitute XS-3™, 1 gallon

CAS Number:  
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
XYLENE SUBSTITUTE XS-3 1GAL Catalog #76548-059

Product 76548-059 is a replacement for 76548-058 but varies by Brand, Packaging or Quantity.

Product Details & Documents

100% aliphatic hydrocarbon for processing and staining.

  • Higher permissible exposure limit than xylene
  • Virtually no odor
  • Flash point 104°F
  • Exceptionally high purity

Part of the StatLab line of clearing agents.

Manufactured in the USA

Specifications

Specifications

  • Description:
    Xylene Substitute XS-3™
  • Size:
    1 gallon

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.