ContactUsLinkComponent
Bluing Reagent Select, Statlab
  76548-234
 :  StatLab
 :  
undefined
Bluing Reagent Select, Statlab
  76548-234
 :  StatLab
 :  SL403
 :  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    Bluing Reagent Select
  • Size:
    1 gallon
  • Cat. No.:
    76548-234
A buffered alkaline rinse with a mild pH (8.0).
  • Ensures optimal staining and proper cellular hue staining
  • Premixed and ready- to-use
  • Tinted blue for easy identification
Part of the StatLab Select stain line.
Manufactured in USA.