- Description:Bluing Reagent Vintage
- Size:1 gallon
- Cat. No.:76548-210
A buffered alkaline rinse with a mild pH (8.0).
- Ensures optimal staining and proper cellular contrast
- Premixed and ready-to-use.
Part of the StatLab Vintage stain line.
Manufactured in USA.