Rat Insulin-like Growth Factor 2 mRNA-Binding Protein 2 (IGF2BP2) ELISA Kit, AFG Bioscience
76576-670
:
- Assay Type (ELISA with LOV):Sandwich
- Host:Rat
- Target protein:Insulin-like growth factor 2 mRNA-binding protein 2
- Description:Rat insulin-like growth factor 2 mRNA-binding protein 2 (IGF2BP2) ELISA kit
- Size:96 Wells
- Sample Type:Serum, plasma, urine, tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:-
- Storage Temperature:2…8 °C
- Sample Volume:50 - 100 µl
- Sensitivity:-
- Regulatory Status:RUO
- Cat. No.:76576-670
- Research topic: Epigenetics and nuclear signaling, signal transduction
- Abbreviation: IGF2BP2
- Detection wavelength: 450 nm