Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

α,3,5-Tribromotoluene 99%

Glass bottle for solids, 5 g
Catalog # TS37978-0050
Corrosive
Flammable
 :  37978-0050
α,3,5-Tribromotoluene 99%
Catalog # TS37978-0050
CAS Number:  56908-88-4
α,3,5-Tribromotoluene 99%
α,3,5-Tribromotoluene 99%

Glass bottle for solids, 5 g

CAS Number:  56908-88-4
Price
$189.00
Log in for availability
Each (5g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  3,5-Dibromobenzyl bromide, 1,3-Dibromo-5-(bromomethyl)benzene, alpha,3,5-Tribromotoluene
  • CAS Number:56908-88-4
  • Formula:C₇H₅Br₃
  • MDL Number:MFCD00052415
  • Molecular Weight:328.83
  • Boiling Pt:173 °C (19 mmHg)
  • Melting Pt:93…97 °C
  • UN:2921
  • ADR:8,II
Specifications

Specification Test Results

  • Appearance:
    White to beige to light brown Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    93 to 97 °C
  • GC:
    ≥98.5 %

Specifications

  • Pack type:
    Glass bottle for solids
  • Size:
    5 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.